Learn More
TADA2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11299-1-AP-20UL
 View more versions of this product
Product Code. 16888273
  • £135.00 / 20µL
Explore more special offers
Description
Description
TADA2L (transcriptional adapter 2-like), also known as TADA2A (transcriptional adapter 2-alpha) or ADA2-like protein, is a 443 amino acid nuclear protein that exists as 2 alternatively spliced isoforms. While most abundantly expressed in testis, TADA2L is present in all tissues. TADA2L contains one SANT domain and one SWIRM domain, and interacts with GCN5 and GR (NR3C1). Its ability to bind double-stranded DNA allows TADA2L to play a role in chromatin remodeling. Although it makes up part of the PCAF complex, TADA2L is also a component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4. The gene that encodes TADA2L contains 71,408 bases and maps to human chromosome 17q12. Chromosome 7 houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome.
Specifications
Specifications
TADA2L
Polyclonal
Unconjugated
TADA2A
ADA2, ADA2 like protein, ADA2A, hADA2, KL04P, TADA2A, TADA2L, Transcriptional adapter 2 like
Rabbit
Antigen Affinity Chromatography
RUO
217031, 360581, 6871
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75478, Q6AYE3, Q8CHV6
Tada2a
TADA2L Fusion Protein Ag1817
20 μL
Primary
Rat, Mouse, Human