missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24235-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the TET family of RNA-binding proteins. The encoded protein plays a role in RNA polymerase II gene transcription as a component of a distinct subset of multi-subunit transcription initiation factor TFIID complexes. Translocations involving this gene play a role in acute leukemia and extraskeletal myxoid chondrosarcoma, and mutations in this gene may play a role in amyotrophic lateral sclerosis. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
TAF15 | |
Polyclonal | |
Unconjugated | |
TAF15 | |
hTAFII68, RBP56, RNA binding protein 56, TAF(II)68, TAF15, TAF2N, TAFII68 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8148 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92804 | |
TAF15 | |
TAF15 Fusion Protein Ag20828 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |