missing translation for 'onlineSavingsMsg'
Learn More

TAF15 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TAF15
Concentration 0.43 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16882735
View Documents
Proteintech
24235-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16872735
View Documents
Proteintech
24235-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a member of the TET family of RNA-binding proteins. The encoded protein plays a role in RNA polymerase II gene transcription as a component of a distinct subset of multi-subunit transcription initiation factor TFIID complexes. Translocations involving this gene play a role in acute leukemia and extraskeletal myxoid chondrosarcoma, and mutations in this gene may play a role in amyotrophic lateral sclerosis. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

TAF15
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q92804
8148
TAF15 Fusion Protein Ag20828
Primary
-20°C
TAF15
0.43 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
hTAFII68, RBP56, RNA binding protein 56, TAF(II)68, TAF15, TAF2N, TAFII68
TAF15
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications