missing translation for 'onlineSavingsMsg'
Learn More

TAF1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55170-1-AP-20UL

 View more versions of this product

Product Code. 16816655

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibody is specific to TAF1L.

TAF1L, also named as TAF (II)210, belongs to the TAF1 family. It may act as a functional substitute for TAF1/TAFII250 during male meiosis, when sex chromosomes are transcriptionally silenced.
Specifications

Specifications

TAF1L
Polyclonal
Unconjugated
TAF1L
TAF(II)210, TAF1L, TAF2A2, TBP associated factor 1 like, TBP associated factor 210 kDa
Rabbit
Antigen Affinity Chromatography
RUO
138474
-20°C
Liquid
Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IZX4
TAF1L
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers