Learn More
Tocris Bioscience™ Talampanel
Non-competitive AMPA/kainate antagonist
Brand:  Tocris Bioscience™ 5032/10
Product Code. 18705537
  • £168.00 / 10mg
Explore more special offers
Description
Description
Talampanel is a non-competitive AMPA/kainate receptor antagonist that displays 2.3-3-fold more potent activity than GYKI 52466 (Cat. No. 1454). Potentiates the anticonvulsive activity of antiepileptic drugs in animal models of seizures. Orally active.
Specifications
Specifications
>98%
Talampanel
Talampanel
10 mg
337.37