missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
25 kg
Packaging:
Plastic Bottle
Grade:
Reagent Pharmacopeia Chemistry Reagents
Description
- Leading the industry for over 65 years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 800°C |
CAS | 14807-96-6 |
Color | White |
pH | 8.2 to 8.5 (20°C) (Suspension) |
Quantity | 25 kg |
Molecular Formula | H2Mg3O12Si4 |
Synonym | Hydrous magnesium silicate |
InChI Key | KNXVOGGZOFOROK-UHFFFAOYSA-N |
SMILES | [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
IUPAC Name | trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'