missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, particle size: -200 mesh (<75 μm), Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 800.0°C |
Color | White |
Physical Form | Very Fine Powder |
Quantity | 10 kg |
Linear Formula | Mg3[Si4O10(OH)2] |
Merck Index | 15,9168 |
Solubility Information | Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkalies |
Formula Weight | 379.28 |
Concentration or Composition (by Analyte or Components) | Typical chemical properties: SiO2: 59.3%, MgO: 31.5%, Al2O3: 1.9%, Fe2O3: 1.0%, CaO: 0.6% |
Loss on Drying | 0.2% (105°C) max. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'