Learn More
Invitrogen™ Tali™ Cellular Analysis Slides
The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer.
Brand: Invitrogen™ T10794
261.25 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 μL of sample per chamber. Each box contains 50 slides.
Ready to Use for Your Convenience
Each Tali™ Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali™ Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.
Order Info
Shipping Condition: Room temperature
Specifications
Cellular Analysis Slide | |
Tali™ Image-Based Cytometer | |
Contains one box of 50 slides. Store at room temperature. | |
Room Temperature |
Tali | |
Fluorescence | |
50 Slides |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.