missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14168-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
Talin-1 | |
Polyclonal | |
Unconjugated | |
TLN1 | |
ILWEQ, KIAA1027, talin 1, TLN, TLN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21894, 313494, 7094 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26039, Q9Y490 | |
Tln1 | |
Talin-1 Fusion Protein Ag5367 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |