Learn More
Köttermann Tall Safety Storage Cabinet with Doors
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
£4059.80 - £4732.90
Product Code | Brand | Door Style | No. of Shelves | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Door Style | No. of Shelves | Includes | Price | Quantity & Availability | |||
17382129
|
Köttermann
371-00031 |
Hinged, Auto-close | None | 5 Internal Drawers |
£4642.80
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17362129
|
Köttermann
371-00013 |
Hinged, Auto-close | 3 | 1 Drip Tray |
£4059.80
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17372129
|
Köttermann
371-00030 |
Hinged, Auto-close | 6 | 1 Drip Tray |
£4181.70
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17312139
|
Köttermann
371-00035 |
Hinged, Auto-close | 4 | 1 Drip Tray |
£4112.80
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17302139
|
Köttermann
371-00034 |
Folding, Manual-close | None | 4 Internal Drawers |
£4732.90
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Different versions that meet your needs
- Additional storage space
- Top safety for flammable hazardous substances (Type 90)
Specifications
615 x 1200 x 1955 mm | |
2 |