missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva TALON™ Superflow Medium
Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.
£191.00 - £672.00
Specifications
For Use With (Application) | For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography |
---|---|
Storage Requirements | 4°C to 8°C |
Binding Capacity | Up to 20 mg Histidine-tagged Protein |
Product Type | Talon Superflow Media |
Particle Size | 60 to 160 μm |
Description
Features:
- Ligand - TALON Superflow Cobalt-IMAC
- Matrix - Cross-linked agarose, 6%
- Particle Size - 60 -160μm
- Binding Capacity/mL - Up to 20 mg histidine-tagged protein
- pH stability working range - 3 to 12
- pH stability Cleaning-in-Place (CIP) - 2 - 14 Co2+stripped medium
- Complete packsize - 10mL
Specifications
For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography | |
Up to 20 mg Histidine-tagged Protein | |
60 to 160 μm | |
3 to 12 | |
Talon Superflow Cobalt-IMAC |
4°C to 8°C | |
Talon Superflow Media | |
6% Cross-Linked Agarose | |
20% Ethanol |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title