missing translation for 'onlineSavingsMsg'
Learn More
Tamoxifen Free Base MP Biomedicals Product Code.: 11435161
compliance-icons

Tamoxifen Free Base MP Biomedicals

Product Code. 11435161
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 11435161

Brand: MP Biomedicals 0215673880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 10540-29-1
Molecular Formula C26H29NO
MDL Number MFCD00010454
Quantity 100 mg
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Molecular Weight (g/mol) 371.52
PubChem CID 2733526
ChEBI CHEBI:41774
Show More Show Less
compliance-icons
Product Identifier
  • Tamoxifen, Free Base
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tamoxifen Free Base MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.