Learn More
Corning™ Tank Liners
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
£68.35 - £108.42
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Product Type | Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Product Type | Size | Price | Quantity & Availability | ||
17203594
|
Corning™
91-300-30 |
200 L | 57.15 cm | Non-Gusseted Tank Liner | 22.5 in. x 36 in. (57.2 cm x 91.4 cm) |
£108.42
Each |
Estimated Shipment: 11-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17213594
|
Corning™
91-300-35 |
200 L | 55.88 cm | Gusseted Tank Liner | 22 in. x 36 in. (55.9 cm x 91.4 cm) |
£68.35
Each |
Estimated Shipment: 11-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Made from low-density polyethylene (LDPE)
- Eliminate costs associated with cleaning and validation of tanks
- Decrease labor costs and increase turnaround time
- Eliminate risks associated with cross contamination
- Help extend life of reuseable tanks, resulting in reduced capital expenditures
- Gamma irradiated, individually packaged
- Choice of gusseted and non-gusseted available
- Available in many sizes
Specifications
LDPE | |
Cylindrical |