Learn More
Fisherbrand™ Tank Vent Filter
Contains activated carbon. Fisher Scientific™ Tank Vent Filter also provides membrane filtration and chemical absorption.
Brand:  Fisherbrand™
Additional Details : Weight : 0.16200kg
Product Code. 15409689
  • £167.00 / Each
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
Description
Description
  • Prevents airborne particles, organic and bacteria from entering the PE water tank.
  • Integral scavenger removes CO2
Specifications
Specifications
Accu500
Filter, Vent
CO2 scavenger