missing translation for 'onlineSavingsMsg'
Learn More
Tannic acid, 95%, Thermo Scientific Chemicals Product Code.: 10484745

Tannic acid, 95%, Thermo Scientific Chemicals

Product Code. 10484745
5 kg, Plastic drum
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10484745

Brand: Thermo Scientific Acros 202420051

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Melting Point 218.0°C
Color White to Yellow
Assay Percent Range 95%
Merck Index 15, 9184
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: soluble in glycerol, ethanol and acetone;, practically insoluble in benzene, chcl3, diethyl e, ther, petroleum ether, cs2, ccl4, hexane;, slightly soluble in anhydrous ethanol.
Formula Weight 1701.23
Percent Purity 95%
Density 198g/mL
Quantity 5 kg
Flash Point 198°C
Infrared Spectrum Authentic
Loss on Drying 12.0% max.
Packaging Plastic drum
Physical Form Beads
Chemical Name or Material Tannic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tannic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.