missing translation for 'onlineSavingsMsg'
Learn More

Tannic Acid, Honeywell Fluka™
BIOPHARMA

Product Code. 15661540
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
25 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss., meets analytical specification of USP, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

UN Number NONH for all modes of transport
Beilstein 8186396
Solubility Information Soluble in acetone, alcohol, water 1gm/0.35mL, glycerol 1gm/1mL (warm). Insoluble in carbon disulfide, chloroform, diethyl ether,carbon tetrachloride, petroleum ether,benzene, hexane. Slightly soluble in dehydrated alcohol.
Formula Weight 1701.2g/mol
Grade puriss.
Quantity 1 kg
Flash Point 199°C (390.2°F)
Ignition Point 980°F
Packaging Plastic Bottle
Physical Form Powder
Residue after Ignition ≤0.1% (as SO4)
Chemical Name or Material Tannic Acid, Meets analytical specification of USP
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'