missing translation for 'onlineSavingsMsg'
Learn More

Tanshinone IIA, MedChemExpress

MedChemExpress Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.

£68.97 - £251.68

Specifications

Chemical Name or Material Tanshinone IIA
CAS 568-72-9
Color Red
Purity Grade Notes Research
Molecular Formula C19H18O3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16493836
Encompass_Preferred
View Documents
Medchem Express
HY-N0135
25 mg
£136.73
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16444767
Encompass_Preferred
View Documents
Medchem Express
HY-N0135
50 mg
£251.68
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16428878
Encompass_Preferred
View Documents
Medchem Express
HY-N0135
10 mg
£68.97
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Specifications

Specifications

Tanshinone IIA
Red
C19H18O3
DMSO : 100 mg/mL (339.74 mM; Need ultrasonic)
294.34
99.17%
Solid
568-72-9
Research
Dan Shen ketone
O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O
294.34
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.