missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum disilicide, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Tantalum disilicide is used in the preparation of ceramics, which finds application in the thermal protection systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 2200°C |
Physical Form | Powder |
Quantity | 10 g |
Solubility Information | Insoluble in water. |
Formula Weight | 237.12 |
Chemical Name or Material | Tantalum disilicide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'