missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 0.127mm (0.005in) thick, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11305779
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
200 x 200 mm
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]

Form: Approx. 5.3g/50 x 50mm; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray to Silver
Appearance Gray to silver foil
Assay Percent Range 99.95% (Metals basis)
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Chemical Name or Material Tantalum, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'