missing translation for 'onlineSavingsMsg'
Learn More
Tantalum foil, 2.0mm (0.08in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11345779

Tantalum foil, 2.0mm (0.08in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11345779
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy

Product Code. 11345779

Brand: Thermo Scientific Alfa Aesar 010357.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.95%
Odor Odorless
Weight ≈20.8g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Tantalum foil, 2.0mm (0.08 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L013 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number010357.GH,010357.FF,010357.FI
Lot NumberT06J005 Certificate TypeCertificate of Analysis Date26/07/2023 Catalog Number010357.GH,010357.FF,010357.FI
Lot NumberZ30I015 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog Number010357.GH,010357.FF,010357.FI
Lot NumberU01I012 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number010357.GH,010357.FF,010357.FI
Lot NumberU01I011 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number010357.GH,010357.FF,010357.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum foil, 2.0mm (0.08in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.