missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
POA - £101.51
Chemical Identifiers
CAS | 7440-25-7 |
---|---|
Molecular Formula | Ta |
Molecular Weight (g/mol) | 180.95 |
MDL Number | MFCD00011252 |
InChI Key | GUVRBAGPIYLISA-UHFFFAOYSA-N |
Synonym | tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod |
PubChem CID | 23956 |
ChEBI | CHEBI:33348 |
IUPAC Name | tantalum |
SMILES | [Ta] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15487285
|
Thermo Scientific Alfa Aesar
046695.FL |
75 x 75 mm |
£101.51
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15497285
|
Thermo Scientific Alfa Aesar
046695.GT |
150 x 150 mm | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-25-7 | |
180.95 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
23956 | |
tantalum |
Ta | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
CHEBI:33348 | |
[Ta] |
Specifications
7440-25-7 | |
MFCD00011252 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
tantalum | |
23956 | |
100 Mesh | |
Wire Cloth |
Ta | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
[Ta] | |
180.95 | |
CHEBI:33348 | |
Odorless | |
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave |
Safety and Handling
missing translation for 'einecsNumber' : 231-13505
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only