missing translation for 'onlineSavingsMsg'
Learn More
Tantalum pellet, 1mm (0.04in) dia x 2mm (0.08in), 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15404915

Tantalum pellet, 1mm (0.04in) dia x 2mm (0.08in), 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15404915
1000 Pc.
Click to view available options
Quantity:
1000 Pc.
This item is not returnable. View return policy

Product Code. 15404915

Brand: Thermo Scientific Alfa Aesar 044367.KO

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-25-7
Molecular Formula Ta
MDL Number MFCD00011252
Quantity 1000 Pc.
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES [Ta]
IUPAC Name tantalum
Molecular Weight (g/mol) 180.95
PubChem CID 23956
ChEBI CHEBI:33348
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tantalum pellet, 1mm (0.04 in.) dia. x 2mm (0.08in)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25H007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044367.KO
Lot NumberJ13P31 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044367.KO
Lot NumberD26R031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044367.KO
Lot NumberP21F005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044367.KO
Lot NumberX04D014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044367.KO

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum pellet, 1mm (0.04in) dia x 2mm (0.08in), 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.