missing translation for 'onlineSavingsMsg'
Learn More
Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals Product Code.: 11336847

Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals

Product Code. 11336847
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11336847

Brand: Thermo Scientific Alfa Aesar 010753.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Black to Gray
Appearance Gray to black powder
Assay Percent Range 99.98% (Metals basis, Nb 50ppm)
Quantity 25 g
Mesh Size -22 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Tantalum

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY02J018 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number010753.06,010753.14,010753.22
Lot NumberW13J010 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number010753.06,010753.14,010753.22
Lot NumberQ11J008 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number010753.14
Lot NumberP17H011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010756.06,010753.06,010753.14,010753.22
Lot Number22279 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010756.06,010753.06,010753.14,010753.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.