Learn More
Tantalum powder, -325 mesh, Puratronic™, 99.97% (metals basis), Nb 50ppm, Thermo Scientific Chemicals
£27.94 - £1211.03
Chemical Identifiers
CAS | 7440-25-7 |
---|---|
Molecular Formula | Ta |
Molecular Weight (g/mol) | 180.95 |
MDL Number | MFCD00011252 |
InChI Key | GUVRBAGPIYLISA-UHFFFAOYSA-N |
Synonym | tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod |
PubChem CID | 23956 |
ChEBI | CHEBI:33348 |
IUPAC Name | tantalum |
SMILES | [Ta] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335788
|
Thermo Scientific Alfa Aesar
010345.88 |
2.5 g |
£27.94
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15485935
|
Thermo Scientific Alfa Aesar
010345.14 |
25 g |
£117.59
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355788
|
Thermo Scientific Alfa Aesar
010345.22 |
100 g |
£325.62
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325788
|
Thermo Scientific Alfa Aesar
010345.36 |
500 g |
£1211.03
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-25-7 | |
180.95 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
23956 | |
tantalum |
Ta | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rodShow More | |
CHEBI:33348 | |
[Ta] |
Specifications
Black to Gray | |
Gray to black powder | |
Ta | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
[Ta] | |
180.95 | |
CHEBI:33348 | |
Odorless | |
Tantalum |
7440-25-7 | |
99.97% (Metals basis, Nb 50ppm) | |
MFCD00011252 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
tantalum | |
23956 | |
-325 Mesh | |
Powder |
Safety and Handling
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-135-5
missing translation for 'rtecsNumber' : WW5505000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.