missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, APS ^=2 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11374647
Click to view available options
Quantity:
5 g
25 g
50 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Black
UN Number UN3089
Quantity 50 g
Percent Purity 99.9%
Odor Odorless
Particle Size APS ≈2 Micron
Form Powder
Assay (metals basis)
Chemical Name or Material Tantalum powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'