missing translation for 'onlineSavingsMsg'
Learn More

Tantalum rod, 12.7mm (0.5in) dia, hard, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11305798
Click to view available options
Quantity:
1 cm
5 cm
25 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Quantity 25 cm
Percent Purity 99.95%
Odor Odorless
Weight ≈20.4g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Tantalum rod, 12.7mm (0.5 in.) dia., Hard, Temper: as drawn

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'