missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) bromide, 99.9% (metals basis)

Product Code. 11357657
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
This item is not returnable. View return policy

Product Code. 11357657

Brand: Thermo Scientific Alfa Aesar 014041.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13451-11-1 | Br5Ta | 580.468 g/mol

The Tantalum Bromide Cluster is used for the preparation of heavy-atom derivatives for structure determination of biological macromolecules by X-ray analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-11-1
Molecular Formula Br5Ta
Molecular Weight (g/mol) 580.468
MDL Number MFCD00049563
InChI Key GCPVYIPZZUPXPB-UHFFFAOYSA-I
Synonym tantalum pentabromide, tantalum bromide tabr5, tantalum v bromide, tantalum 5+ pentabromide, tabr5
PubChem CID 83480
IUPAC Name pentabromotantalum
SMILES Br[Ta](Br)(Br)(Br)Br

Specifications

Boiling Point 348.8°C
Melting Point 265°C
Color Yellow
Physical Form Powder
Quantity 10 g
UN Number UN3260
Solubility Information Insoluble in water.
Formula Weight 580.49
Percent Purity 99.9%
Odor Pungent
Sensitivity Moisture sensitive
Density 4.67 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum(V) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.