missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10721481

Tantalum(V) chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10721481
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10721481

Brand: Thermo Scientific Acros 203910050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242.0°C
Melting Point 221.0°C
Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 99.99%
Linear Formula TaCl5
UN Number 3260
Fieser 16,312; 17,321
Merck Index 15, 9186
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in alcohol, koh, h2so4
Formula Weight 358.2
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Tantalum(V) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467235 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number203910050,203910250,203911000
Lot NumberA0454143 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number203910050,203910250,203911000
Lot NumberA0447775 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number203910050,203910250,203911000
Lot NumberA0415456 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number203910050,203910250,203911000
Lot NumberA0438135 Certificate TypeCertificate of Analysis Date17/01/2022 Catalog Number203910050,203910250,203911000
compliance-icons compliance-icons
Product Identifier
  • Tantalum(V) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.