missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11387018

Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11387018
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11387018

Brand: Thermo Scientific Alfa Aesar 014708.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242°C
Melting Point 216°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
UN Number UN3260
Merck Index 14,9054
Solubility Information Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide. Slightly soluble in ethanol.
Formula Weight 358.21
Percent Purity 99.99%
Grade Puratronic™
Odor Acidic
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.68 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum(V) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT03F004 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number014708.06,014708.22,014708.14
Lot NumberU07K009 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number014708.06,014708.22,014708.14
Lot NumberU11J019 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number014708.06,014708.22,014708.14
Lot NumberT25E216 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number014708.06,014708.22,014708.14
Lot NumberE28S998 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014708.06,014708.22,014708.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.