missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) ethoxide, optical grade, 99.95% (metal basis), Thermo Scientific Chemicals Product Code.: 11302877

Tantalum(V) ethoxide, optical grade, 99.95% (metal basis), Thermo Scientific Chemicals

Product Code. 11302877
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11302877

Brand: Thermo Scientific Alfa Aesar L16033.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6074-84-6
Molecular Formula C10H25O5Ta
Molecular Weight (g/mol) 406.25
MDL Number MFCD00049785
InChI Key HSXKFDGTKKAEHL-UHFFFAOYSA-N
Synonym tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
PubChem CID 160806
IUPAC Name ethanolate;tantalum(5+)
SMILES CCO[Ta](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 145°C (0.1mmHg)
Melting Point 18°C to 19°C
Quantity 5 g
Linear Formula Ta(OCH2CH3)5
UN Number UN2924
Beilstein 3678999
Flash Point 30°C (86°F)
Formula Weight 406.25
Percent Purity 99.95%
Grade Optical
Refractive Index 1.488
Sensitivity Air and moisture sensitive
Density 1.566
Chemical Name or Material Tantalum(V) ethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ30G005 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberL16033.03,L16033.06
Lot NumberZ16G013 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog NumberL16033.03,L16033.06
Lot Number10138481 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16033.03,L16033.06
Lot NumberM02F014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16033.03,L16033.06
Lot Number10176160 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16033.03,L16033.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) ethoxide, optical grade, 99.95% (metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.