missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm, Thermo Scientific Chemicals Product Code.: 11333428

Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm, Thermo Scientific Chemicals

Product Code. 11333428
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11333428

Brand: Thermo Scientific Alfa Aesar 036343.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tantalum (V) oxide (Ta2O5) has potential applications as part of an environmental barrier coating system for Si3N4-based turbine components. Octadecyl phosphoric acid ester has been found to produce oriented, well-ordered monolayers on a flat tantalum (V) oxide surface, via self-assembly from a heptane/propan-2-ol solution. Tantalum(V) oxide (Ta 2 O 5 ) has been used extensively as an insulating layer in discrete capacitors and shows a high potential for semiconductor applications such as an insulating dielectric in the capacitive element in memory devices such as dynamic random access.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Sintered Lump
Quantity 25 g
Merck Index 14,9056
Solubility Information Insoluble in water.
Formula Weight 441.89
Percent Purity 99.95%
Odor Odorless
Density 8.2 g/mL
Assay (metals basis excluding Nb), Nb <200ppm
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252549 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog Number036343.22,036343.14
Lot NumberP03I023 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036343.22,036343.14
Lot NumberC19I10 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036343.22,036343.14
Lot NumberJ30R048 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036343.22,036343.14
Lot NumberD25K16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036343.22,036343.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.