Learn More
Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals
Description
Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1800°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Insoluble in water,alcohol,mineral acids. Soluble in HFInsoluble in almost all organic solvents including alcohol and mineral acids. Solubility is negligible in water. Soluble in HF |
Formula Weight | 441.89 |
Density | 8.2 |
Chemical Name or Material | Tantalum(V) oxide |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0451091 | Certificate of Analysis | 20/03/2024 | 014709.09,014709.18,014709.30 |
T11J013 | Certificate of Analysis | 18/07/2023 | 014709.09,014709.18,014709.30 |
M20I017 | Certificate of Analysis | 12/04/2022 | 014709.09,014709.18,014709.30,014713.AE |
M20I018 | Certificate of Analysis | 12/04/2022 | 014709.09,014709.18,014709.30,014713.AE |
W18B043 | Certificate of Analysis | 07/12/2021 | 014709.09,014709.18,014709.30,014713.AE |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.