missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11317028

Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11317028
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11317028

Brand: Thermo Scientific Alfa Aesar 014709.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
Synonym Tantalum pentoxide
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Powder
Quantity 250 g
Merck Index 14,9056
Solubility Information Insoluble in water,alcohol,mineral acids; Soluble in HFInsoluble in almost all organic solvents including alcohol and mineral acids; Solubility is negligible in water; Soluble in HF
Formula Weight 441.89
Percent Purity 99%
Odor Odorless
Density 8.2
Assay (metals basis)
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451091 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number014709.09,014709.18,014709.30
Lot NumberT11J013 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number014709.09,014709.18,014709.30
Lot NumberM20I017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014709.09,014709.18,014709.30,014713.AE
Lot NumberM20I018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014709.09,014709.18,014709.30,014713.AE
Lot NumberW18B043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014709.09,014709.18,014709.30,014713.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) oxide, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.