missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, Puratronic™, 99.993% (metals basis excluding Nb), Nb 50ppm max

Product Code. p-7048563
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11347057

Brand: Thermo Scientific Alfa Aesar 010881.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-61-0 | O5Ta2 | 441.89 g/mol

It is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components, and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits, and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Powder
Quantity 25 g
Merck Index 14,9056
Solubility Information Insoluble in almost all organic solvents. Solubility is negligible in water.
Formula Weight 441.89
Percent Purity 99.993%
Grade Puratronic
Odor Odorless
Density 8.2 g/mL
Assay (metals basis excluding Nb), Nb 50ppm max.
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.