missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, Spectrographic Grade, 99.5% (metals basis), Thermo Scientific Chemicals

£71.86 - £203.01

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344879
View Documents
Thermo Scientific Alfa Aesar
073118.14
25 g
£71.86
25g
Please to purchase this item. Need a web account? Register with us today!
11354879
View Documents
Thermo Scientific Alfa Aesar
073118.22
100 g
£203.01
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components, and electronic circuitry. In addition, tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits, and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-61-0
441.89
PBCFLUZVCVVTBY-UHFFFAOYSA-N
O=[Ta](=O)O[Ta](=O)=O
O5Ta2
MFCD00011254
[(dioxotantalio)oxy]dioxotantalum

Specifications

1800°C
Powder
MFCD00011254
Insoluble in almost all organic solvents. Solubility is negligible in water.
O=[Ta](=O)O[Ta](=O)=O
441.89
99.5%
Odorless
(metals basis)
1314-61-0
O5Ta2
14,9056
PBCFLUZVCVVTBY-UHFFFAOYSA-N
[(dioxotantalio)oxy]dioxotantalum
441.89
Spectrographic
8.2 g/mL
Tantalum (V) oxide
Videos
Safety and Handling

Safety and Handling

H500

missing translation for 'einecsNumber' : 215-238-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only