missing translation for 'onlineSavingsMsg'
Learn More
Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11355798

Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11355798
25 m
Click to view available options
Quantity:
1 m
25 m
5 m
This item is not returnable. View return policy

Product Code. 11355798

Brand: Thermo Scientific Alfa Aesar 010349.G9

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 3.26g/m; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Gray to Silver
Appearance Gray to silver wire
Density 16.69g/cm3
Assay Percent Range 99.95% (Metals basis)
Quantity 25 m
Odor Odorless
Form Wire
Chemical Name or Material Tantalum, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX31J032 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number010349.G1,010349.G5,010349.G9
Lot NumberX31J033 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number010349.G1,010349.G5,010349.G9
Lot NumberS03I011 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number010349.G1,010349.G5,010349.G9
Lot NumberC22X005 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010349.G1,010349.G5,010349.G9
Lot NumberU24E038 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010349.G1,010349.G5,010349.G9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.