missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAOK1 Recombinant Human Protein
TAOK1 is a serine/threonine-protein kinase. Recombinant human TAOK1 (1-314) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6416
This item is not returnable.
View return policy
Description
TAOK1 is involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
62.1kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 1-314 | |
Recombinant |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol | |
100μg | |
PSK2, MARKK, MAP3K16, FLJ14314, KIAA1361 | |
Human | |
GST Tag |