missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Silicone Stopcock Grease

Provides excellent heat and cold resistance without considerable change of consistency. Karl Hecht™ Assistent™ Silicone Stopcock Grease is a medium viscosity grease.

Brand:  Karl Hecht™ 41480035

Product Code. 10671152

  • £11.15 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tubes of 35g
  • Consistency between -50° to +150°C
Specifications

Specifications

Tap Grease
Silicone
35 g
Medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers