missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TAP Growth Media, optimized for Chlamydomonas culture
GREENER_CHOICE

Product Code. p-7076275
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
6000mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12393553

Brand: Gibco™ A1379801

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

TAP Growth Media, optimized for Chlamydomonas culture

Gibco™ TAP Media is optimized for the growth and maintenance of Chlamydomonas reinhardtii. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.

Each Gibco™ TAP Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helping you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Chlamydomonas culture, results in better overall efficiency and more robust data (Fig. 1, 2).

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Growth Medium
Quantity 1 L
Shipping Condition Room Temperature
Preparation Method None (Ready-to-Use)
Species C. reinhardtii
Content And Storage Gibco™ TAP Growth Media contains:

•  1 L Gibco™ TAP Media — Store at 4 °C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.