missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Tape Dispenser

This Tape Dispenser dispenses the tape quickly, safely and efficiently. Tape is simple to advance by using your thumb and steel safety cutting blade cuts fast and smooth.

Brand:  Brady™ 072412

Product Code. 11751925

  • £136.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This tape dispenser holds 305 m long rolls of tape with a 75 mm width.
Specifications

Specifications

Tape Dispenser
4 in.
101.6 x 254 x 304.80 mm
254 mm
304.8 mm
Component and equipment labelling, General Identification, Inventory and inspection labelling, Lean and 5S labelling
Black
101.6 mm
10 in.
12 in.
Tape Dispenser - holds rolls up to 305 m long, with max. tape width of 75 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers