missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq Core Kit 10

Supplied with standard reaction buffers

Brand:  MP Biomedicals™ 11EPTQK105C

Code : 80

Additional Details : Weight : 0.17500kg

 View more versions of this product

Product Code. 11437030

  • £895.70 / 1250 units
Estimated Shipment: 11-09-2024
to see stock.

Includes: Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix

Description

Description

Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.

Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
Specifications

Specifications

Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C
Frozen liquid
Standard PCR
Frozen
5 x 250U
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only (RUO)