missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq DNA Pol

A thermostable DNA polymerase that is used for PCR, RT-PCR, DNA sequencing and DNA labeling.

Brand:  MP Biomedicals™ 11EPTQD025

 View more versions of this product

Product Code. 11487020

  • £160.00 / 250 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5' to 3' polymerase and exonuclease activities.

Unit Definition: One unit will incorporate 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.

Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5′'to 3' polymerase and exonuclease activities.
Unit Definition: One unit will incorporate 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C
Liquid
Standard PCR, RT-PCR, Sequencing, Nucleic Acid Labeling
Taq DNA Pol
250U
5U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.