missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq Pol Bulk

Product Code. p-8015249 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 250U
1000U
250U
3 x 250U
5 x 1000U
Unit Size:
1000 units
250 units
750 units
This item is not returnable. View return policy

Product Code. 11467010

Brand: MP Biomedicals™ 11EPTQA025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For life science applications

  • Ready for CE IVD labeling of clinical applications
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Description Taq Pol Bulk
Quantity 250U
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

For Resaerch Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.