Learn More
TaqMan™ TaqMan™ Advanced miRNA cDNA Synthesis Kit
Kit uses 3' poly-A tailing and 5' ligation of an adaptor sequence to extend the mature miRNAs present in the sample on each end prior to reverse transcription.
Brand: TaqMan™ A28007
213.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The Applied Biosystems TaqMan Advanced miRNA cDNA Synthesis Kit uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA Assays for detection and quantification of mature miRNAs in biological samples. The kit is designed to perform 50 cDNA synthesis reactions, with theresulting cDNA sufficient for up to 600 qPCR reactions, allowing extensive downstream analysis.
Features of the TaqMan Advanced miRNA workflow include:• Sensitivity: high sensitivity allows for quantification of low-expressing miRNA targets, especially in biological fluids such as plasma, serum, and tissue
• Flexibility: the universal RT chemistry generates universal cDNA templates for all miRNAs present in the sample, allowing for flexibility in study design
• Sample savings: requires only 2 μL of plasma/serum sample input and generates enough cDNA for 600 qPCR reactions (20 μL) or 1200 qPCR reactions (10 μL)
• Specificity: 5’ ligation step allows for single base pair discrimination along entire miRNA sequence, including difficult-to-detect mismatches on 5’ end
• Gold-standard TaqMan chemistry: This kit is designed for use with TaqMan Advanced miRNA Assays for qPCR, which draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and validation system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families.
The TaqMan Advanced miRNA cDNA Synthesis Kit uses 3' poly-A tailing and 5' ligation of an adaptor sequence to extend the mature miRNAs present in the sample on each end prior to reverse transcription. Universal RT primers recognize the universal sequences present on both the 5' and 3' extended ends of the mature miRNAs. All mature miRNAs in the sample are reverse transcribed to cDNA.
In order to improve detection of low-expressing miRNA targets, the cDNA is then amplified using the Universal miR-Amp Primers and miR-Amp Master Mix to uniformly increase the amount of cDNA for each target, maintaining the relative differential expression levels. Unlike traditional pre-amplification, the Universal miR-Amp Primers recognize the universal sequences added to all mature miRNAs on the 5' and 3' ends, ensuring that there is no amplification bias.
Note: the TaqMan Advanced miRNA cDNA Synthesis Kit is not compatible with first generation TaqMan MicroRNA Assays for qPCR. For reverse transcription of miRNA targets using miRNA-specific RT primers, please see the TaqMan MicroRNA Reverse Transcription Kit.
Order Info
Note: the TaqMan Advanced miRNA cDNA Synthesis Kit is not compatible with first generation TaqMan MicroRNA Assays for qPCR. For reverse transcription of miRNA targets using miRNA-specific RT primers. Shipping Conditions: Dry Ice

Specifications
• 10X PolyA Buffer • ATP, 10mM • Poly A Enzyme, 5 U/μL • 5X Ligase Buffer • RNA Ligase, 10 U/μL • 50% PEG 8000 • 25X Ligation Adaptor • 10X RT Enzyme Mix • 20X Universal RT Primer • dNTP Mix, 100mM • 20X miR-Amp Primer Mix • 5X RT Buffer • 2X miR-Amp Master Mix Store all at -5 to -30°C, except 2X miR-Amp Master Mix which is stored at 2–8°C. |
|
High | |
Standard | |
42°C | |
cDNA Synthesis Kit | |
SuperScript™ III | |
miRNA analysis | |
Separate Components | |
10 μL |
Primer-probe | |
qPCR | |
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex | |
TaqMan™ | |
50 cDNA synthesis reactions | |
Dry Ice | |
50 cDNA synthesis reactions / 600 qPCR reactions | |
RNA |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.