missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Fast, 96-well Plate; Format 96
TaqMan™ Array, Fast, 96-well Plate; Format 96
Brand: Applied Biosystems™ 4413255
167.14 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Custom TaqMan™ Plates are cost-effective and convenient 96-well formats for TaqMan™ Gene Expression Assays. Choose from over 50,000 inventoried human, mouse, rat, canine, and rhesus macaque assays, and from five plate formats - single assays, or assays in duplicate or triplicate.Specifications
8 x 12; 96-well Plate; Format 96 | |
Custom TaqMan™ Array Fast Plates, 10 μL rxns, 96-Well Plate Format, 1 Sample and 95 Controls in TaqMan™ Array Fast Plates for TaqMan™ Gene Expression Assays | |
Fast | |
1 plate | |
10 μL |
Store at room temperature. | |
Standard | |
TaqMan™ | |
Gene Expression, Genotyping |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction