missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Format 16
TaqMan™ Array, Format 16
Brand: Applied Biosystems™ 4346798
Code : NEW
Additional Details : Weight : 0.00010kg
Description
The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.Specifications
Store in refrigerator (2°C to 8°C) | |
TaqMan™ | |
1 Ea. |
16 | |
Custom Gene Expression TaqMan™ Array Cards |
For Research Use Only. Not for use in diagnostic procedures.