Learn More
Applied Biosystems™ TaqMan™ Array, Human Angiogenesis, Fast 96-well
TaqMan™ Array, Human Angiogenesis, Fast 96-well
Brand: Applied Biosystems™ 4418716
Code : 50
Additional Details : Weight : 0.00450kg
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human Angiogenesis Plate targets known angiogenesis growth factors like VEGF as well as matrix derived inhibitors such as endostatin. Additionally, the panel contains markers and targets for angiogenesis and lymphangiogenesis.Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
1 x 10 μL Reactions |
Dried Down | |
Fast | |
FAM | |
Angiogenesis, Oncology | |
Gene Expression, Oncology, Genetic Disease Research |
For Research Use Only. Not for use in diagnostic procedures.