missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Human eNOS Signaling, Fast 96-well
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
1 set
Beschreibung
- These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state
- Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment
- The TaqMan™ Array Human eNOS Signaling 96-well Plate contains:
- 92 assays to genes associated with eNOS signaling
- 4 assays to candidate endogenous control genes
Spezifikation
Spezifikation
| Type | Array Plate |
| Format | 96-array Plate |
| Species | Human |
| Content And Storage | Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. |
| Form | Dried Down |
| Includes | TaqMan Assay for One 10 uL Reaction |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan™ |
| Quantity | 1 x 96-well plate |
| Mehr anzeigen |
Dokumentation
Product Information
Specification Sheets
Product Bulletin: TaqMan Array Plates
File size: 1 MB
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
Applied Biosystems™ TaqMan™ Array, Human eNOS Signaling, Fast 96-well >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur