Learn More
Applied Biosystems™ TaqMan™ Array, Human Hedgehog, Fast 96-well
TaqMan™ Array, Human Hedgehog, Fast 96-well
Brand: Applied Biosystems™ 4418734
Code : 50
Additional Details : Weight : 0.00450kg
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human Hedgehog Pathway Plate targets genes from the three members of the vertebrate hedgehog (hh) family: Desert hh, Indian hh, and Sonic hh. Also included are two Patched homologs,Ptc1 and Ptc2; and three homologs of Ci (Cubitus interruptus); Gli1,Gli2 and Gli3. Genes from two cell-cell signaling pathways which commonly function in association with Hedgehog signaling (WNT and TGFB) are also included. Other genes involved in signal transduction of the hedgehog signals (MAPK⁄ERK) as well as genes associated with cell differentiation and regulation of development are included.Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
1 x 10 μL Reactions |
Dried Down | |
Fast | |
FAM | |
Cell Cycle & Proliferation, Apoptosis, Neurobiology, Oncology, Development, Immunology, Signal Transduction | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.