Learn More
Applied Biosystems™ TaqMan™ Array, Human Immune, Fast 96-well
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state.
Brand: Applied Biosystems™ 4418718
119.56 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human Immune Response Plate targets genes from immune system functions that fall into 9 classes: Cell Surface Receptors; Stress Response; Oxidoreductases; Proteases; Transcription Factors; Signal Transduction; Cytokines & Cytokine Receptors; Chemokines & Chemokine Receptors; and Cell Cycle & Protein Kinases.
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
1 x 10 μL Reactions |
Dried Down | |
Fast | |
FAM | |
Cell Cycle & Proliferation, Signal Transduction, Apoptosis, Neurobiology | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.